Gene Transcripts
- Gene Symbol
- RAE1
- Entrez Gene ID
- 8480
- Gene Name
- ribonucleic acid export 1
- Gene Aliases
- MIG14, MRNP41, Mnrp41, dJ481F12.3, dJ800J21.1
- Chromosome Location
- Chr. 20: 57350969 - 57378463 on Build GRCh38
- UniGene ID
- Hs.371698
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK055170.1 |
- |
1 |
2169 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
R96431.1 |
- |
|
269 |
|
Chr. - - [-] |
View Full Product Details