Gene Transcripts

Gene Symbol
LCORL
Entrez Gene ID
254251
Gene Name
ligand dependent nuclear receptor corepressor like
Gene Aliases
MLR1
Chromosome Location
Chr. 4: 17841199 - 18023287 on Build GRCh38
UniGene ID
Hs.446201
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153686.8 NP_710153.2 7 4758
NR_136669.1 5 4584
XM_011513821.2 XP_011512123.1 9 9489
XM_011513822.2 XP_011512124.1 8 9462
XM_011513823.2 XP_011512125.1 8 9335
XM_011513826.2 XP_011512128.1 6 4542
XM_017007961.1 XP_016863450.1 8 10771
XM_017007962.1 XP_016863451.1 8 9413
XM_017007963.1 XP_016863452.1 8 9716
GenBank mRNA AL133031.1 - 1 2887

View Full Product Details