Gene Transcripts

Gene Symbol
ITPKB
Entrez Gene ID
3707
Gene Name
inositol-trisphosphate 3-kinase B
Gene Aliases
IP3-3KB, IP3K, IP3K-B, IP3KB, PIG37
Chromosome Location
Chr. 1: 226631690 - 226739327 on Build GRCh38
UniGene ID
Hs.528087
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017001211.1 XP_016856700.1 3 9286
GenBank mRNA BC086865.1 - 1 1776

View Full Product Details