Gene Transcripts
- Gene Symbol
- SPAG11B
- Entrez Gene ID
- 10407
- Gene Name
- sperm associated antigen 11B
- Gene Aliases
- EDDM2B, EP2, EP2C, EP2D, HE2, HE2C, SPAG11
- Chromosome Location
- Chr. 8: 7447754 - 7463670 on Build GRCh38
- UniGene ID
- Hs.2717
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL133652.1 |
- |
1 |
1894 |
|
View Full Product Details