Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001184763.1 | NP_001171692.1 | 8 | 6248 | |
NM_144624.2 | NP_653225.2 | 7 | 6500 | ||
NM_175866.4 | NP_787062.1 | 8 | 6589 | ||
GenBank mRNA | AK026668.1 | - | 1 | 436 | |
AL137257.1 | - | 1 | 2598 | ||
BC001977.1 | - | 1 | 1321 | ||
BC043436.2 | - | 1 | 2314 |