Gene Transcripts
- Gene Symbol
- FARP2
- Entrez Gene ID
- 9855
- Gene Name
- FERM, ARH/RhoGEF and pleckstrin domain protein 2
- Gene Aliases
- FIR, FRG, PLEKHC3
- Chromosome Location
- Chr. 2: 241356249 - 241494842 on Build GRCh38
- UniGene ID
- Hs.726316
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC036887.1 |
- |
2 |
1829 |
|
View Full Product Details