Gene Transcripts
- Gene Symbol
- ANK2
- Entrez Gene ID
- 287
- Gene Name
- ankyrin 2, neuronal
- Gene Aliases
- ANK-2, LQT4, brank-2
- Chromosome Location
- Chr. 4: 112706083 - 113383740 on Build GRCh38
- UniGene ID
- Hs.620557
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK128049.1 |
- |
1 |
32 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB062981.1 |
- |
1 |
32 |
|
Chr. 4 - 113369786 - 113370325 [+] |
GenBankmRNA |
DB062992.1 |
- |
1 |
32 |
|
Chr. 4 - 113369786 - 113370328 [+] |
View Full Product Details