Gene Transcripts
- Gene Symbol
- DCAF12
- Entrez Gene ID
- 25853
- Gene Name
- DDB1 and CUL4 associated factor 12
- Gene Aliases
- CT102, KIAA1892, TCC52, WDR40A
- Chromosome Location
- Chr. 9: 34086383 - 34126773 on Build GRCh38
- UniGene ID
- Hs.493750
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF085907.1 |
- |
1 |
495 |
|
View Full Product Details