Gene Transcripts
- Gene Symbol
- TRG-AS1
- Entrez Gene ID
- 100506776
- Gene Name
- T cell receptor gamma locus antisense RNA 1
- Gene Aliases
- TCRGV
- Chromosome Location
- Chr. 7: 38341577 - 38378637 on Build GRCh38
- UniGene ID
- Hs.173034
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK130649.1 |
- |
1 |
972 |
|
View Full Product Details