Gene Transcripts
- Gene Symbol
- SYNE1
- Entrez Gene ID
- 23345
- Gene Name
- spectrin repeat containing nuclear envelope protein 1
- Gene Aliases
- 8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
- Chromosome Location
- Chr. 6: 152121684 - 152637399 on Build GRCh38
- UniGene ID
- Hs.12967
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
KU310547.1 |
- |
20 |
4731 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG619667.1 |
- |
1 |
260 |
|
Chr. 6 - 152433702 - 152434485 [-] |
GenBankmRNA |
BC031033.1 |
- |
1 |
260 |
|
Chr. 6 - 152433525 - 152434485 [-] |
View Full Product Details