Gene Transcripts
- Gene Symbol
- AGBL4
- Entrez Gene ID
- 84871
- Gene Name
- ATP/GTP binding protein like 4
- Gene Aliases
- CCP6
- Chromosome Location
- Chr. 1: 48520157 - 50024011 on Build GRCh38
- UniGene ID
- Hs.551844
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF075049.1 |
- |
1 |
392 |
|
View Full Product Details