Gene Transcripts
- Gene Symbol
- PES1
- Entrez Gene ID
- 23481
- Gene Name
- pescadillo ribosomal biogenesis factor 1
- Gene Aliases
- PES
- Chromosome Location
- Chr. 22: 30576625 - 30607013 on Build GRCh38
- UniGene ID
- Hs.517543
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125778.1 |
- |
1 |
373 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB060169.1 |
- |
1 |
373 |
|
Chr. 22 - 30579613 - 30580074 [-] |
View Full Product Details