Gene Transcripts

Gene Symbol
SLC4A8
Entrez Gene ID
9498
Gene Name
solute carrier family 4 member 8
Gene Aliases
NBC3, NDCBE
Chromosome Location
Chr. 12: 51391317 - 51515763 on Build GRCh38
UniGene ID
Hs.4749
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039960.2 NP_001035049.1 25 9117
NM_001258401.2 NP_001245330.1 25 9063
XM_011539010.1 XP_011537312.1 26 9172
XM_011539011.1 XP_011537313.1 26 9117
XM_011539014.2 XP_011537316.1 16 7770
XM_017020240.1 XP_016875729.1 26 9199
XM_017020242.1 XP_016875731.1 26 9186
GenBank mRNA AK124576.1 - 1 2315
AL137310.1 - 1 1071

View Full Product Details