Gene Transcripts

Gene Symbol
ATP2C1
Entrez Gene ID
27032
Gene Name
ATPase secretory pathway Ca2+ transporting 1
Gene Aliases
ATP2C1A, BCPM, HHD, PMR1, SPCA1, hSPCA1
Chromosome Location
Chr. 3: 130850500 - 131016712 on Build GRCh38
UniGene ID
Hs.584884
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AL833453.1 - 1 805

View Full Product Details