Gene Transcripts
- Gene Symbol
- PAPPA
- Entrez Gene ID
- 5069
- Gene Name
- pappalysin 1
- Gene Aliases
- ASBABP2, DIPLA1, IGFBP-4ase, PAPA, PAPP-A, PAPPA1
- Chromosome Location
- Chr. 9: 116153752 - 116402321 on Build GRCh38
- UniGene ID
- Hs.643599
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK024330.1 |
- |
1 |
21 |
|
View Full Product Details