Gene Transcripts

Gene Symbol
WDR61
Entrez Gene ID
80349
Gene Name
WD repeat domain 61
Gene Aliases
REC14, SKI8
Chromosome Location
Chr. 15: 78283233 - 78299726 on Build GRCh38
UniGene ID
Hs.513055
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK310477.1 - 2 224


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DB086449.1 - 2 224 Chr. 15 - 78295285 - 78299703 [-]
GenBankmRNA AL109709.1 - 1 1848 Chr. 15 - 78295402 - 78297490 [-]

View Full Product Details