Gene Transcripts
- Gene Symbol
- HFE
- Entrez Gene ID
- 3077
- Gene Name
- hemochromatosis
- Gene Aliases
- HFE1, HH, HLA-H, MVCD7, TFQTL2
- Chromosome Location
- Chr. 6: 26087281 - 26096216 on Build GRCh38
- UniGene ID
- Hs.233325
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA957218.1 |
- |
1 |
459 |
|
Chr. 6 - 26095509 - 26096070 [+] |
GenBankmRNA |
BX107211.1 |
- |
1 |
530 |
|
Chr. 6 - 26095471 - 26096083 [+] |
GenBankmRNA |
AA102329.1 |
- |
1 |
212 |
|
Chr. 6 - 26095759 - 26096093 [+] |
View Full Product Details