Gene Transcripts
- Gene Symbol
- FBXL4
- Entrez Gene ID
- 26235
- Gene Name
- F-box and leucine rich repeat protein 4
- Gene Aliases
- FBL4, FBL5, MTDPS13
- Chromosome Location
- Chr. 6: 98868535 - 98948006 on Build GRCh38
- UniGene ID
- Hs.536850
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DN996736.1 |
- |
1 |
355 |
|
Chr. 6 - 98870548 - 98871119 [-] |
GenBankmRNA |
DA816824.1 |
- |
1 |
107 |
|
Chr. 6 - 98870308 - 98870878 [-] |
GenBankmRNA |
W46956.1 |
- |
|
20 |
|
Chr. - - [-] |
View Full Product Details