Gene Transcripts
- Gene Symbol
- DERL2
- Entrez Gene ID
- 51009
- Gene Name
- derlin 2
- Gene Aliases
- CGI-101, DERtrin-2, F-LAN-1, F-LANa, FLANa
- Chromosome Location
- Chr. 17: 5471251 - 5486230 on Build GRCh38
- UniGene ID
- Hs.286131
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CB851413.1 |
- |
1 |
618 |
|
Chr. 17 - 5471324 - 5472060 [-] |
GenBankmRNA |
BM714595.1 |
- |
1 |
193 |
|
Chr. 17 - 5471251 - 5471642 [-] |
GenBankmRNA |
F29564.1 |
- |
1 |
100 |
|
Chr. 17 - 5471264 - 5471549 [-] |
GenBankmRNA |
CN294099.1 |
- |
|
401 |
|
Chr. - - [-] |
GenBankmRNA |
AW292566.1 |
- |
1 |
368 |
|
Chr. 17 - 5471365 - 5471817 [-] |
GenBankmRNA |
DB513231.1 |
- |
1 |
244 |
|
Chr. 17 - 5471258 - 5471693 [-] |
View Full Product Details