Gene Transcripts
- Gene Symbol
- SLC8A1
- Entrez Gene ID
- 6546
- Gene Name
- solute carrier family 8 member A1
- Gene Aliases
- NCX1
- Chromosome Location
- Chr. 2: 40112146 - 40512452 on Build GRCh38
- UniGene ID
- Hs.31961
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK095013.1 |
- |
1 |
1010 |
|
View Full Product Details