Gene Transcripts

Gene Symbol
OR2F1
Entrez Gene ID
26211
Gene Name
olfactory receptor family 2 subfamily F member 1 (gene/pseudogene)
Gene Aliases
7M1-2, OLF3, OR14-60, OR2F3, OR2F3P, OR2F4, OR2F5, OR7-139, OR7-140
Chromosome Location
Chr. 7: 143907152 - 143988540 on Build GRCh38
UniGene ID
Hs.553595
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011516019.2 XP_011514321.1 3 550
XM_011516020.2 XP_011514322.1 3 325
GenBank mRNA AJ459868.1 - 2 225

View Full Product Details