Gene Transcripts

Gene Symbol
PSPN
Entrez Gene ID
5623
Gene Name
persephin
Gene Aliases
PSP
Chromosome Location
Chr. 19: 6375294 - 6375849 on Build GRCh38
UniGene ID
Hs.248159
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK055490.1 - 1 2155

Gene Symbol
ALKBH7
Entrez Gene ID
84266
Gene Name
alkB homolog 7
Gene Aliases
ABH7, SPATA11, UNQ6002
Chromosome Location
Chr. 19: 6372433 - 6375250 on Build GRCh38
UniGene ID
Hs.111099
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BM271861.1 - 1 113

View Full Product Details