Gene Transcripts
- Gene Symbol
- MORN1
- Entrez Gene ID
- 79906
- Gene Name
- MORN repeat containing 1
- Gene Aliases
- Chromosome Location
- Chr. 1: 2321253 - 2391751 on Build GRCh38
- UniGene ID
- Hs.642701
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC029954.1 |
- |
1 |
1352 |
|
View Full Product Details