Gene Transcripts

Gene Symbol
TRAF3IP2-AS1
Entrez Gene ID
643749
Gene Name
TRAF3IP2 antisense RNA 1
Gene Aliases
C6UAS, C6orf3, NCRNA00248, TRAF3IP2-AS2
Chromosome Location
Chr. 6: 111483472 - 111602295 on Build GRCh38
UniGene ID
Hs.486228
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_034108.1 4 4624
NR_034109.1 2 4333
GenBank mRNA AF136408.1 - 1 2102

View Full Product Details