Gene Transcripts
- Gene Symbol
- OXR1
- Entrez Gene ID
- 55074
- Gene Name
- oxidation resistance 1
- Gene Aliases
- Nbla00307, TLDC3
- Chromosome Location
- Chr. 8: 106270178 - 106752694 on Build GRCh38
- UniGene ID
- Hs.127286
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK098168.1 |
- |
1 |
504 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB203805.1 |
- |
1 |
504 |
|
Chr. 8 - 106749730 - 106750292 [+] |
View Full Product Details