Gene Transcripts
- Gene Symbol
- Hist1h3g
- Entrez Gene ID
- 97908
- Gene Name
- histone cluster 1, H3g
- Gene Aliases
- H3.1-221, M32460
- Chromosome Location
- Chr. 13: 23535418 - 23535909 on Build GRCm38
- UniGene ID
- Mm.261657
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M15434.1 |
- |
1 |
395 |
|
View Full Product Details