Gene Transcripts

Gene Symbol
Mir99ahg
Entrez Gene ID
77994
Gene Name
Mir99a and Mirlet7c-1 host gene (non-protein coding)
Gene Aliases
2810055G20Rik, D730043B02Rik
Chromosome Location
Chr. 16: 77329328 - 77558428 on Build GRCm38
UniGene ID
Mm.394688
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK033222.1 - 20


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DV067527.1 - 1 324 Chr. 16 - 77500215 - 77594785 [+]
GenBankmRNA BB624333.1 - 1 17 Chr. 16 - 77500508 - 77594975 [+]
GenBankmRNA BE654839.1 - 1 141 Chr. 16 - 77500384 - 77691323 [+]

View Full Product Details