Gene Transcripts
- Gene Symbol
- Peli3
- Entrez Gene ID
- 240518
- Gene Name
- pellino 3
- Gene Aliases
- 6030441F14Rik, A930011L17, BC028931
- Chromosome Location
- Chr. 19: 4929717 - 4943127 on Build GRCm38
- UniGene ID
- Mm.277580
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC022735.1 |
- |
1 |
486 |
|
BC057046.1 |
- |
1 |
3439 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG297260.1 |
- |
|
485 |
|
Chr. - - [-] |
View Full Product Details