Gene Transcripts

Gene Symbol
Tnrc6b
Entrez Gene ID
213988
Gene Name
trinucleotide repeat containing 6b
Gene Aliases
2700090M07Rik, A730065C02Rik, AI848765, D230019K20Rik
Chromosome Location
Chr. 15: 80711313 - 80941086 on Build GRCm38
UniGene ID
Mm.131328
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK012593.1 - 1 83


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY711738.1 - 1 80 Chr. 15 - 80884353 - 80884727 [+]

View Full Product Details