Gene Transcripts
- Gene Symbol
- Igk-V28
- Entrez Gene ID
- 16114
- Gene Name
- immunoglobulin kappa chain variable 28 (V28)
- Gene Aliases
- AV061724, Igk-Ef1, Igk-Trp, Igk-VSer
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.333124
- Species
- Mus musculus
- Gene Symbol
- Igkc
- Entrez Gene ID
- 16071
- Gene Name
- immunoglobulin kappa constant
- Gene Aliases
- Igk-C
- Chromosome Location
- Chr. 6: 70726435 - 70726754 on Build GRCm38
- UniGene ID
- Mm.333124
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC092090.1 |
- |
|
722 |
|
BC110685.1 |
- |
|
739 |
|
- Gene Symbol
- Igkj1
- Entrez Gene ID
- 110759
- Gene Name
- immunoglobulin kappa joining 1
- Gene Aliases
- Igk-J1
- Chromosome Location
- Chr. 6: 70722562 - 70722599 on Build GRCm38
- UniGene ID
- Mm.333124
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC013496.1 |
- |
|
699 |
|
- Gene Symbol
- Igkv1-117
- Entrez Gene ID
- 16098
- Gene Name
- immunoglobulin kappa variable 1-117
- Gene Aliases
- Cr1, Gm16797, IgK, Igk-V21, V(k)45.1
- Chromosome Location
- Chr. 6: 68121091 - 68121825 on Build GRCm38
- UniGene ID
- Mm.333124
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC092064.1 |
- |
|
745 |
|
- Gene Symbol
- Igkv3-4
- Entrez Gene ID
- 626347
- Gene Name
- immunoglobulin kappa variable 3-4
- Gene Aliases
- Igk-V21-4
- Chromosome Location
- Chr. 6: 70671789 - 70672377 on Build GRCm38
- UniGene ID
- Mm.333124
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC108385.1 |
- |
|
714 |
|
- Gene Symbol
- Igkv4-53
- Entrez Gene ID
- 546213
- Gene Name
- immunoglobulin kappa variable 4-53
- Gene Aliases
- Gm1420, Gm1526, IgVkkh4, kh4
- Chromosome Location
- Chr. 6: 69648824 - 69649354 on Build GRCm38
- UniGene ID
- Mm.305097
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC092251.1 |
- |
|
719 |
|
BC106159.1 |
- |
|
738 |
|
- Gene Symbol
- Igkv4-59
- Entrez Gene ID
- 667683
- Gene Name
- immunoglobulin kappa variable 4-59
- Gene Aliases
- EG667683, Gm8760, kk4
- Chromosome Location
- Chr. 6: 69438218 - 69438741 on Build GRCm38
- UniGene ID
- Mm.333124
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC091738.1 |
- |
|
739 |
|
- Gene Symbol
- Igkv6-23
- Entrez Gene ID
- 637227
- Gene Name
- immunoglobulin kappa variable 6-23
- Gene Aliases
- 19-23, EG637227, Gm7202, IgK
- Chromosome Location
- Chr. 6: 70260409 - 70260933 on Build GRCm38
- UniGene ID
- Mm.305097
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC091754.1 |
- |
|
716 |
|
- Gene Symbol
- Igkv8-16
- Entrez Gene ID
- 640340
- Gene Name
- immunoglobulin kappa variable 8-16
- Gene Aliases
- Igk-V8-16
- Chromosome Location
- Chr. 6: 70386672 - 70387238 on Build GRCm38
- UniGene ID
- Mm.333124
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC082779.1 |
- |
|
724 |
|
- Gene Symbol
- Igkv8-30
- Entrez Gene ID
- 384419
- Gene Name
- immunoglobulin kappa chain variable 8-30
- Gene Aliases
- ENSMUSG00000073025, ENSMUSG00000076577, Gm10883
- Chromosome Location
- Chr. 6: 70117061 - 70117617 on Build GRCm38
- UniGene ID
- Mm.333124
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC094049.1 |
- |
|
727 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BC055911.1 |
- |
|
751 |
3662102 |
Chr. - - [-] |
GenBankmRNA |
BC021781.1 |
- |
|
730 |
4216859 |
Chr. - - [-] |
GenBankmRNA |
BC106172.1 |
- |
|
714 |
|
Chr. - - [-] |
GenBankmRNA |
S65921.1 |
- |
|
786 |
|
Chr. - - [-] |
View Full Product Details