Gene Transcripts
- Gene Symbol
- Slc44a1
- Entrez Gene ID
- 100434
- Gene Name
- solute carrier family 44, member 1
- Gene Aliases
- 2210409B22Rik, 4833416H08Rik, AW547365, CHTL1, CTL1, Cdw92
- Chromosome Location
- Chr. 4: 53440413 - 53622478 on Build GRCm38
- UniGene ID
- Mm.482207
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK008866.1 |
- |
1 |
435 |
|
AK141895.1 |
- |
1 |
1652 |
|
AK151977.1 |
- |
1 |
1181 |
|
View Full Product Details