Gene Transcripts
- Gene Symbol
- Slx1b
- Entrez Gene ID
- 75764
- Gene Name
- SLX1 structure-specific endonuclease subunit homolog B (S. cerevisiae)
- Gene Aliases
- 1110030E23Rik, 2410170E21Rik, 4833422P03Rik, AI853643, Giyd1, Giyd2
- Chromosome Location
- Chr. 7: 126688927 - 126695783 on Build GRCm38
- UniGene ID
- Mm.280573
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AA097433.1 |
- |
1 |
398 |
|
Chr. 7 - 126689525 - 126690047 [-] |
GenBankmRNA |
AI595193.1 |
- |
1 |
399 |
|
Chr. 7 - 126689526 - 126690047 [-] |
View Full Product Details