Gene Transcripts

Gene Symbol
Prox2
Entrez Gene ID
73422
Gene Name
prospero homeobox 2
Gene Aliases
1700058C01Rik
Chromosome Location
Chr. 12: 85085810 - 85110770 on Build GRCm38
UniGene ID
Mm.311944
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK086375.1 - 1 159


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB662944.1 - 1 159 Chr. 12 - 85086374 - 85087707 [+]

View Full Product Details