Gene Transcripts
- Gene Symbol
- Prox2
- Entrez Gene ID
- 73422
- Gene Name
- prospero homeobox 2
- Gene Aliases
- 1700058C01Rik
- Chromosome Location
- Chr. 12: 85085810 - 85110770 on Build GRCm38
- UniGene ID
- Mm.311944
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK086375.1 |
- |
2 |
499 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB662944.1 |
- |
2 |
499 |
|
Chr. 12 - 85086374 - 85087707 [+] |
View Full Product Details