Gene Transcripts
- Gene Symbol
- Arsb
- Entrez Gene ID
- 11881
- Gene Name
- arylsulfatase B
- Gene Aliases
- 1110007C02Rik, AI480648, As-1, As-1r, As-1s, As-1t, As1, As1-r, As1-s, As1-t, Asr-1, Ast-1
- Chromosome Location
- Chr. 13: 93771023 - 93943016 on Build GRCm38
- UniGene ID
- Mm.300178
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK037155.1 |
- |
1 |
930 |
|
View Full Product Details