Gene Transcripts
- Gene Symbol
- Apaf1
- Entrez Gene ID
- 11783
- Gene Name
- apoptotic peptidase activating factor 1
- Gene Aliases
- 6230400I06Rik, Apaf-1, Apaf1l, fog, mKIAA0413
- Chromosome Location
- Chr. 10: 90989311 - 91082743 on Build GRCm38
- UniGene ID
- Mm.220289
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK053432.1 |
- |
1 |
436 |
|
View Full Product Details