Gene Transcripts
- Gene Symbol
- Crnde
- Entrez Gene ID
- 71296
- Gene Name
- colorectal neoplasia differentially expressed (non-protein coding)
- Gene Aliases
- 4933436C20Rik
- Chromosome Location
- Chr. 8: 92326031 - 92356120 on Build GRCm38
- UniGene ID
- Mm.86664
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB751125.1 |
- |
2 |
121 |
|
Chr. 8 - 92326038 - 92344943 [-] |
GenBankmRNA |
AW985587.1 |
- |
3 |
321 |
|
Chr. 8 - 92326186 - 92346453 [-] |
GenBankmRNA |
DT913885.1 |
- |
4 |
418 |
|
Chr. 8 - 92326124 - 92346870 [-] |
View Full Product Details