Gene Transcripts
- Gene Symbol
- Nacad
- Entrez Gene ID
- 192950
- Gene Name
- NAC alpha domain containing
- Gene Aliases
- D230024G13Rik, mKIAA0363
- Chromosome Location
- Chr. 11: 6597814 - 6606054 on Build GRCm38
- UniGene ID
- Mm.337771
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK046108.1 |
- |
1 |
1609 |
|
View Full Product Details