Gene Transcripts
- Gene Symbol
- Tmem86b
- Entrez Gene ID
- 68255
- Gene Name
- transmembrane protein 86B
- Gene Aliases
- C330014O21Rik
- Chromosome Location
- Chr. 7: 4628040 - 4631395 on Build GRCm38
- UniGene ID
- Mm.389784
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020752.1 |
- |
|
339 |
|
View Full Product Details