Gene Transcripts

Gene Symbol
Mbd2
Entrez Gene ID
17191
Gene Name
methyl-CpG binding domain protein 2
Gene Aliases
MBD2a
Chromosome Location
Chr. 18: 70568270 - 70626131 on Build GRCm38
UniGene ID
Mm.322
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF072244.1 - 1 14


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DT922271.1 - 176 Chr. - - [-]

View Full Product Details