Gene Transcripts
- Gene Symbol
- Slc1a3
- Entrez Gene ID
- 20512
- Gene Name
- solute carrier family 1 (glial high affinity glutamate transporter), member 3
- Gene Aliases
- AI504299, B430115D02Rik, Eaat1, GLAST, GLAST-1, GLU-T, GluT-1, Gmt1, MGluT1
- Chromosome Location
- Chr. 15: 8634124 - 8710807 on Build GRCm38
- UniGene ID
- Mm.204834
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK046688.1 |
- |
1 |
2087 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB328290.2 |
- |
1 |
331 |
|
Chr. 15 - 8685833 - 8686495 [-] |
View Full Product Details