Gene Transcripts

Gene Symbol
Ddi2
Entrez Gene ID
68817
Gene Name
DNA-damage inducible protein 2
Gene Aliases
1110056G13Rik, 1700027M01Rik, 9130022E05Rik, AI604911, AU040698
Chromosome Location
Chr. 4: 141683563 - 141723419 on Build GRCm38
UniGene ID
Mm.393721
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK018643.1 - 536
AK041061.1 - 1199
AK078461.1 - 908
AK129015.1 - 5401
AK165215.1 - 297

View Full Product Details