Gene Transcripts
- Gene Symbol
- Ddi2
- Entrez Gene ID
- 68817
- Gene Name
- DNA-damage inducible protein 2
- Gene Aliases
- 1110056G13Rik, 1700027M01Rik, 9130022E05Rik, AI604911, AU040698
- Chromosome Location
- Chr. 4: 141683563 - 141723419 on Build GRCm38
- UniGene ID
- Mm.393721
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK165215.1 |
- |
|
1982 |
|
View Full Product Details