Gene Transcripts
- Gene Symbol
- Uba6
- Entrez Gene ID
- 231380
- Gene Name
- ubiquitin-like modifier activating enzyme 6
- Gene Aliases
- 4930542H01, 5730469D23Rik, AU021846, AW124799, E1-L2, Ube1l2
- Chromosome Location
- Chr. 5: 86110720 - 86172743 on Build GRCm38
- UniGene ID
- Mm.34012
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK017687.1 |
- |
|
921 |
|
AK029740.1 |
- |
|
2205 |
|
AK139327.1 |
- |
|
1363 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ257842.1 |
- |
1 |
680 |
|
Chr. 5 - 86109507 - 86110201 [-] |
View Full Product Details