Gene Transcripts

Gene Symbol
Cbx1
Entrez Gene ID
12412
Gene Name
chromobox 1
Gene Aliases
Cbx, Cbx-rs2, E430007M08Rik, HP1B, Hp1beta, M31, MOD1
Chromosome Location
Chr. 11: 96788679 - 96808982 on Build GRCm38
UniGene ID
Mm.29055
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK080243.1 - 1 497
AK088236.1 - 1 477


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AA474107.1 - 1 241 Chr. 11 - 96807458 - 96807748 [+]
GenBankmRNA AI227435.1 - 1 407 Chr. 11 - 96807291 - 96807738 [+]
GenBankmRNA AA671074.1 - 1 208 Chr. 11 - 96807491 - 96807925 [+]
GenBankmRNA BY762162.1 - 1 542 Chr. 11 - 96807251 - 96808147 [+]
GenBankmRNA AA591657.1 - 1 276 Chr. 11 - 96807423 - 96807712 [+]
GenBankmRNA BY746258.1 - 477 Chr. - - [-]
GenBankmRNA BE648625.1 - 1 318 Chr. 11 - 96807380 - 96808081 [+]
GenBankmRNA AV573079.1 - 1 198 Chr. 11 - 96807501 - 96807737 [+]
GenBankmRNA BE850431.1 - 1 242 Chr. 11 - 96807456 - 96807938 [+]
GenBankmRNA BB637650.1 - 1 497 Chr. 11 - 96807201 - 96807817 [+]
GenBankmRNA CD563934.1 - 1 195 Chr. 11 - 96807503 - 96807767 [+]
GenBankmRNA CK621202.1 - 1 469 Chr. 11 - 96807229 - 96807761 [+]
GenBankmRNA CK790554.1 - 1 463 Chr. 11 - 96807251 - 96807959 [+]
GenBankmRNA AI158488.1 - 1 290 Chr. 11 - 96807408 - 96807799 [+]

View Full Product Details