Gene Transcripts
- Gene Symbol
- Lrrc8d
- Entrez Gene ID
- 231549
- Gene Name
- leucine rich repeat containing 8D
- Gene Aliases
- 2810473G09Rik, 4930525N13Rik, A930019F03, Lrrc5
- Chromosome Location
- Chr. 5: 105699969 - 105815215 on Build GRCm38
- UniGene ID
- Mm.23837
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK013401.1 |
- |
1 |
670 |
|
AK170452.1 |
- |
2 |
2845 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY712394.1 |
- |
1 |
666 |
|
Chr. 5 - 105730678 - 105731477 [+] |
View Full Product Details