Gene Transcripts
- Gene Symbol
- Nrip1
- Entrez Gene ID
- 268903
- Gene Name
- nuclear receptor interacting protein 1
- Gene Aliases
- 6030458L20Rik, 8430438I05Rik, 9630050P12, AA959574, AW456757, RIP140
- Chromosome Location
- Chr. 16: 76287403 - 76374195 on Build GRCm38
- UniGene ID
- Mm.447018
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK078829.1 |
- |
1 |
655 |
|
AK088511.1 |
- |
1 |
2895 |
|
BC062260.1 |
- |
1 |
346 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CK031677.1 |
- |
1 |
671 |
|
Chr. 16 - 76287934 - 76288662 [-] |
View Full Product Details