Gene Transcripts
- Gene Symbol
- Phactr4
- Entrez Gene ID
- 100169
- Gene Name
- phosphatase and actin regulator 4
- Gene Aliases
- 3110001B12Rik, AI527228, AW495572, C330013F19Rik, N28169, mKIAA4120
- Chromosome Location
- Chr. 4: 132355922 - 132422446 on Build GRCm38
- UniGene ID
- Mm.158361
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK013935.1 |
- |
1 |
102 |
|
View Full Product Details