Gene Transcripts
- Gene Symbol
- Atf7
- Entrez Gene ID
- 223922
- Gene Name
- activating transcription factor 7
- Gene Aliases
- 1110012F10Rik, 9430065F09Rik, AI549878, C130020M04Rik
- Chromosome Location
- Chr. 15: 102536643 - 102625421 on Build GRCm38
- UniGene ID
- Mm.208714
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK003630.1 |
- |
|
1272 |
|
AK170250.1 |
- |
|
1362 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ109070.1 |
- |
1 |
22 |
|
Chr. 15 - 102528641 - 102529081 [-] |
View Full Product Details