Gene Transcripts
- Gene Symbol
- Adck1
- Entrez Gene ID
- 72113
- Gene Name
- aarF domain containing kinase 1
- Gene Aliases
- 2610005A10Rik
- Chromosome Location
- Chr. 12: 88360491 - 88461726 on Build GRCm38
- UniGene ID
- Mm.181387
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U01092.1 |
- |
|
305 |
|
- Gene Symbol
- Igk-V1
- Entrez Gene ID
- 16081
- Gene Name
- immunoglobulin kappa chain variable 1 (V1)
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.304143
- Species
- Mus musculus
- Gene Symbol
- Igk-V28
- Entrez Gene ID
- 16114
- Gene Name
- immunoglobulin kappa chain variable 28 (V28)
- Gene Aliases
- AV061724, Igk-Ef1, Igk-Trp, Igk-VSer
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.333124
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC031349.1 |
- |
|
373 |
|
V00810.1 |
- |
2 |
401 |
|
- Gene Symbol
- Igk-V8
- Entrez Gene ID
- 384422
- Gene Name
- immunoglobulin kappa chain variable 8 (V8)
- Gene Aliases
- B128L, B55L, IgK
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.333124
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
D29668.1 |
- |
3 |
305 |
|
U28969.1 |
- |
|
311 |
|
- Gene Symbol
- Igkc
- Entrez Gene ID
- 16071
- Gene Name
- immunoglobulin kappa constant
- Gene Aliases
- Igk-C
- Chromosome Location
- Chr. 6: 70726435 - 70726754 on Build GRCm38
- UniGene ID
- Mm.333124
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC110685.1 |
- |
|
409 |
|
- Gene Symbol
- Igkv1-110
- Entrez Gene ID
- 381777
- Gene Name
- immunoglobulin kappa variable 1-110
- Gene Aliases
- Gm16634, Igk, K5.1
- Chromosome Location
- Chr. 6: 68270527 - 68271265 on Build GRCm38
- UniGene ID
- Mm.333124
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M20828.1 |
- |
|
406 |
|
- Gene Symbol
- Igkv1-117
- Entrez Gene ID
- 16098
- Gene Name
- immunoglobulin kappa variable 1-117
- Gene Aliases
- Cr1, Gm16797, IgK, Igk-V21, V(k)45.1
- Chromosome Location
- Chr. 6: 68121091 - 68121825 on Build GRCm38
- UniGene ID
- Mm.333124
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M55313.1 |
- |
|
416 |
|
- Gene Symbol
- Igkv1-133
- Entrez Gene ID
- 628027
- Gene Name
- immunoglobulin kappa variable 1-133
- Gene Aliases
- EG628027, Gm6827, Igk-Vk2(70/1)
- Chromosome Location
- Chr. 6: 67724914 - 67725661 on Build GRCm38
- UniGene ID
- Mm.333124
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
Z17401.1 |
- |
|
398 |
|
- Gene Symbol
- Igkv1-135
- Entrez Gene ID
- 243420
- Gene Name
- immunoglobulin kappa variable 1-135
- Gene Aliases
- Gm4964, bd2
- Chromosome Location
- Chr. 6: 67609745 - 67610508 on Build GRCm38
- UniGene ID
- Mm.333124
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M17722.1 |
- |
|
380 |
|
M91696.1 |
- |
|
346 |
|
- Gene Symbol
- Igkv14-126
- Entrez Gene ID
- 628127
- Gene Name
- immunoglobulin kappa variable 14-126
- Gene Aliases
- EG628127, Gm6844, IgK, IgM, br9, mga5th128, mga5th137
- Chromosome Location
- Chr. 6: 67896177 - 67896642 on Build GRCm38
- UniGene ID
- Mm.333111
- Species
- Mus musculus
- Gene Symbol
- Igkv16-104
- Entrez Gene ID
- 381778
- Gene Name
- immunoglobulin kappa variable 16-104
- Gene Aliases
- Gm1067, rf
- Chromosome Location
- Chr. 6: 68425605 - 68426072 on Build GRCm38
- UniGene ID
- Mm.333118
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M19911.1 |
- |
|
403 |
|
- Gene Symbol
- Igkv19-93
- Entrez Gene ID
- 692161
- Gene Name
- immunoglobulin kappa chain variable 19-93
- Gene Aliases
- Igk-V38, Vx38C
- Chromosome Location
- Chr. 6: 68736297 - 68736764 on Build GRCm38
- UniGene ID
- Mm.288753
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M17782.1 |
- |
|
365 |
|
- Gene Symbol
- Igkv3-4
- Entrez Gene ID
- 626347
- Gene Name
- immunoglobulin kappa variable 3-4
- Gene Aliases
- Igk-V21-4
- Chromosome Location
- Chr. 6: 70671789 - 70672377 on Build GRCm38
- UniGene ID
- Mm.333124
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC108385.1 |
- |
|
384 |
|
- Gene Symbol
- Igkv4-59
- Entrez Gene ID
- 667683
- Gene Name
- immunoglobulin kappa variable 4-59
- Gene Aliases
- EG667683, Gm8760, kk4
- Chromosome Location
- Chr. 6: 69438218 - 69438741 on Build GRCm38
- UniGene ID
- Mm.333124
- Species
- Mus musculus
- Gene Symbol
- Igkv4-62
- Entrez Gene ID
- 636752
- Gene Name
- immunoglobulin kappa variable 4-62
- Gene Aliases
- EG636752, Gm7188, ab4
- Chromosome Location
- Chr. 6: 69399812 - 69400344 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DQ372804.1 |
- |
|
365 |
|
- Gene Symbol
- Igkv4-68
- Entrez Gene ID
- 384515
- Gene Name
- immunoglobulin kappa variable 4-68
- Gene Aliases
- Gm1419, Gm151, Gm636, IgVk, aa4
- Chromosome Location
- Chr. 6: 69304834 - 69305363 on Build GRCm38
- UniGene ID
- Mm.333129
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DQ372750.1 |
- |
|
364 |
|
- Gene Symbol
- Igkv4-72
- Entrez Gene ID
- 385109
- Gene Name
- immunoglobulin kappa chain variable 4-72
- Gene Aliases
- Gm1499, am4
- Chromosome Location
- Chr. 6: 69226854 - 69227383 on Build GRCm38
- UniGene ID
- Mm.305352
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF178620.1 |
- |
|
308 |
|
AF206030.1 |
- |
|
283 |
|
AY860429.1 |
- |
|
308 |
|
- Gene Symbol
- Igkv4-86
- Entrez Gene ID
- 243451
- Gene Name
- immunoglobulin kappa variable 4-86
- Gene Aliases
- Gm459, kb4
- Chromosome Location
- Chr. 6: 68910411 - 68910938 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DQ372755.1 |
- |
|
359 |
|
- Gene Symbol
- Igkv4-91
- Entrez Gene ID
- 434033
- Gene Name
- immunoglobulin kappa chain variable 4-91
- Gene Aliases
- ENSMUSG00000056850, ENSMUSG00000076532, Gm10879, kf4
- Chromosome Location
- Chr. 6: 68768555 - 68769103 on Build GRCm38
- UniGene ID
- Mm.422999
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF206028.1 |
- |
|
294 |
|
- Gene Symbol
- Igkv5-43
- Entrez Gene ID
- 381783
- Gene Name
- immunoglobulin kappa chain variable 5-43
- Gene Aliases
- Igk-V23, Igkv5-45
- Chromosome Location
- Chr. 6: 69823355 - 69823911 on Build GRCm38
- UniGene ID
- Mm.423821
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF139845.1 |
- |
|
311 |
|
- Gene Symbol
- Igkv6-14
- Entrez Gene ID
- 667881
- Gene Name
- immunoglobulin kappa variable 6-14
- Gene Aliases
- Igk-V19-14
- Chromosome Location
- Chr. 6: 70434952 - 70435476 on Build GRCm38
- UniGene ID
- Mm.426557
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U59155.1 |
- |
|
311 |
|
- Gene Symbol
- Igkv6-20
- Entrez Gene ID
- 108024
- Gene Name
- immunoglobulin kappa variable 6-20
- Gene Aliases
- Igk-V19-20
- Chromosome Location
- Chr. 6: 70335841 - 70336479 on Build GRCm38
- UniGene ID
- Mm.300786
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
L41881.1 |
- |
|
371 |
|
- Gene Symbol
- Igkv6-23
- Entrez Gene ID
- 637227
- Gene Name
- immunoglobulin kappa variable 6-23
- Gene Aliases
- 19-23, EG637227, Gm7202, IgK
- Chromosome Location
- Chr. 6: 70260409 - 70260933 on Build GRCm38
- UniGene ID
- Mm.305097
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF045512.1 |
- |
|
371 |
|
X78108.1 |
- |
|
377 |
|
- Gene Symbol
- Igkv9-120
- Entrez Gene ID
- 434025
- Gene Name
- immunoglobulin kappa chain variable 9-120
- Gene Aliases
- EG434025, Gm5571
- Chromosome Location
- Chr. 6: 68049983 - 68050454 on Build GRCm38
- UniGene ID
- Mm.304143
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF124721.1 |
- |
|
371 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DQ372797.1 |
- |
|
365 |
|
Chr. - - [-] |
GenBankmRNA |
BG961850.1 |
- |
|
391 |
|
Chr. - - [-] |
GenBankmRNA |
FR688609.1 |
- |
|
292 |
|
Chr. - - [-] |
GenBankmRNA |
AY208321.1 |
- |
|
311 |
|
Chr. - - [-] |
GenBankmRNA |
BC021781.1 |
- |
|
400 |
4216859 |
Chr. - - [-] |
GenBankmRNA |
U21578.1 |
- |
|
288 |
|
Chr. - - [-] |
GenBankmRNA |
L08215.1 |
- |
|
308 |
|
Chr. - - [-] |
GenBankmRNA |
AF163747.1 |
- |
|
317 |
|
Chr. - - [-] |
GenBankmRNA |
BE306691.1 |
- |
|
369 |
|
Chr. - - [-] |
GenBankmRNA |
FR689230.1 |
- |
|
142 |
|
Chr. - - [-] |
View Full Product Details